Select a Size
£673.00
Select a Size
About This Item
£673.00
Recommended Products
Joint
joint: 10/600
manufacturer/tradename
Cytiva 18-1153-19
18-1153-13
General description
- High performance glass columns designed for reliable high resolution, reproducibility and durability
- New longer adaptor for increased adjustment of bed height
- Modern, integrated design – simple to assemble, pack, operate, and maintain
- Broad range of column dimensions – suitable for many different purification needs
- Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
- Locking ring – prevents accidental compression of media bed
- Separate packing connector – allows simple column packing for a broad range of media
Choose from one of the most recent versions:
Certificates of Analysis (COA)