Skip to Content
Merck
All Photos(1)

Key Documents

G8523

Sigma-Aldrich

Monoclonal Anti-Growth Hormone antibody produced in mouse

clone GH-C2, ascites fluid

Synonym(s):

Anti-hGH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

clone

GH-C2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
radioimmunoassay: 1:1,200

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

Gene Information

Specificity

The antibody reacts specifically against both natural and recombinant hGH in RIA and ELISA. It shows no cross-reactivity to LH, TSH, glucagon, HCG, HPL, insulin, and prolactin.

Immunogen

recombinant hGH

Physical form

Supplied as diluted ascites fluid containing 1% bovine serum albumin and 15 mM sodium azide.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service