Skip to Content
Merck
All Photos(1)

Key Documents

F3514

Supelco

2-Fluorophenyl 2-nitrophenyl ether

Synonym(s):

2-Fluoro-2′-nitrodiphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4OC6H4NO2
CAS Number:
Molecular Weight:
233.20
Beilstein:
4457734
MDL number:
UNSPSC Code:
12000000

density

1.315 g/mL at 20 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1Oc2ccccc2F

InChI

1S/C12H8FNO3/c13-9-5-1-3-7-11(9)17-12-8-4-2-6-10(12)14(15)16/h1-8H

InChI key

XVZDYNSMGNIZSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service