Skip to Content
Merck
All Photos(1)

Key Documents

F3401

Supelco

Fluorescent Molecular Weight Standards for SDS-PAGE and Protein Transfer Low Range: M.W. 6,500-39,800

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

lyophilized powder

color

blue

storage temp.

−20°C

Other Notes

This product contains the following proteins:
  • Aprotinin, Bovine Lung: 6,500 Da
  • α-Lactalbumin, Bovine Milk: 14,200 Da
  • Trypsin Inhibitor, Soybean: 20,100 Da
  • Carbonic Anhydrase, Bovine Erythrocyte: 29,000 Da
  • Alcohol Dehydrogenase, Horse Liver: 39,800 Da

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service