Key Documents
EP952010115
Adapter for Eppendorf® UVette®, for photometer/spectrophotometer with light path height
size 15 mm
About This Item
UNSPSC Code:
41121815
Product EP952010115 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
Eppendorf® 952010115
size
15 mm
Related Categories
General description
This Eppendorf adaptor is for use with photometers/spectrophotometers which provides 10mm light centered height specifically for the Eppendorf UVette Disposable Cuvette (sold separately). The UVette is ideal for use with the Eppendorf BioPhotometer. In other photometers and/or spectrophotometers, the UVette is inserted with the aid of an adapter. This adapter aligns the UVette to the corresponding height of the optical light path in the instrument. Adapters are also available for use with other common spectrophotometers. Additional adapters for light center heights of 8mm or 15mm are also available (952010107 for 8mm and 952010115 for 15mm). An adapter list is available that provides you with an overview of the correct adapter for use with Eppendorf′s UVette in devices from other manufacturers. No adapter is required for use in Eppendorf′s photometers. The adapters′ compatibility with the devices from other manufacturers has been jointly tested by Eppendorf, the device manufacturers and our customers. Size: 10mm.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)