Skip to Content
Merck
All Photos(3)

Key Documents

EP6335000020

Eppendorf® Mastercycler® Nexus Thermal Cyclers

Flat, complete unit, aluminum block, AC/DC input 115 V, 50/60 Hz (US)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
UNSPSC Code:
41102421

feature

CSA and CE compliant
Flat
aluminum block
complete unit
holds 4 slides or other matching consumables
unit can control 2 other units in network

manufacturer/tradename

Eppendorf® 6335000020 (US)

parameter

37-110 °C temp. range (Lid)
4-99 °C temp. range (control range)

technique(s)

PCR: suitable

W × D × H

25 cm × 41.2 cm × 32.1 cm

weight

10.4 kg

Looking for similar products? Visit Product Comparison Guide

General description

Product Information
With the versatile Mastercycler nexus you can choose the PCR consumables that best suit your assay. The PCR cycler nexus can accommodate 96-well PCR plates, 0.2 mL PCR tubes, 0.2 mL PCR tube strips and 0.5 mL PCR tubes. With a noise level of less than 40 dB[a], the nexus is very quiet and energy-efficient at the same time.

In the age of networking, the new Mastercycler nexus is your reliable companion when it comes to the daily routine of PCR. It is easy to use, does not need much space or energy and sends you an e-mail when it is done – what else do you need from a PCR cycler?

Certified Quality
All Eppendorf cyclers follow these quality guidelines:


  • Individual, documented quality control certificates
  • Calibration accordingly to national and international standards: NIST (USA), DKD/PTB (Germany), UKAS/NPL (UK)
  • UL/cUL listed

The Mastercycler nexus is a whisper quiet PCR machine. Low noise levels make working in your lab more pleasant and enjoyable, particularly when you are thinking about your next big breakthrough findings….

Application

Standard PCR, Fast PCR, PCR optimization

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service