EP5810748003
Eppendorf® Adapter
holds 16 x 3-15ml tubes/ D = 16mm, for rotosr A-4-81 and S-4x500, set of 2 pieces
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
holds 16 x 3-15ml tubes/ D = 16mm, for rotosr A-4-81 and S-4x500
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810748003
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL reactangular bucket
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service