Skip to Content
Merck
All Photos(1)

Key Documents

EP022510525

Eppendorf® PCR-Cooler, 0.2 mL

blue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025
Product EP022510525 is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Eppendorf® 022510525

technique(s)

PCR: suitable

color

blue

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service