EP0030118650
Eppendorf® Conical Tubes SnapTec 50
Eppendorf quality, non-sterile, capacity 45 mL, pack of 8 × 30 ea
Synonym(s):
Conical Centrifuge tube, micro test tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
colorless polypropylene
high-density polyethylene cap (push fit)
sterility
non-sterile
feature
DNase free
packaging
pack of 8 × 30 ea
manufacturer/tradename
Eppendorf® 0030118650
capacity
45 mL
diam.
30 mm
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
SAMPLE STORAGE
Features and Benefits
Premium USP class 6 raw material and no use of slip agents, plasticizers, and biocides during manufacturing ensure highest sample integrity, high g-Safe® centrifugation stability, and optimal sample and pellet visibility
Cap with flattened and grooved sides supports a user-friendly ergonomic handling, enables a stable upright positioning, and minimizes contamination risk
Eppendorf Conical Tubes SnapTec 50 for fast opening and closing in routine multistep protocols
New! Conical Tubes SnapTec 50 are autoclavable at 121°C for 20 Minutes
Reliable and safe labelling due to a large writing area on the tube and due to a flat and light-colored cap
High g-Safe centrifugation stability up to 17,000 x g
Precise dimensions enable maximum compatibility with centrifuge rotors, mixers and shakers
Optimal sample and pellet visibility
Sterile, pyrogen-, DNase-, RNase-, and DNA-free. Each individual lot is checked and certified for quality and purity
Cap with flattened and grooved sides supports a user-friendly ergonomic handling, enables a stable upright positioning, and minimizes contamination risk
Eppendorf Conical Tubes SnapTec 50 for fast opening and closing in routine multistep protocols
New! Conical Tubes SnapTec 50 are autoclavable at 121°C for 20 Minutes
Reliable and safe labelling due to a large writing area on the tube and due to a flat and light-colored cap
High g-Safe centrifugation stability up to 17,000 x g
Precise dimensions enable maximum compatibility with centrifuge rotors, mixers and shakers
Optimal sample and pellet visibility
Sterile, pyrogen-, DNase-, RNase-, and DNA-free. Each individual lot is checked and certified for quality and purity
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service