Skip to Content
Merck
All Photos(1)

Key Documents

EHU051131

Sigma-Aldrich

MISSION® esiRNA

targeting human TGFBR1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
£188.00
50 μG
£334.00

£188.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
£188.00
50 μG
£334.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

£188.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAAGTCATCACCTGGCCTTGGTCCTGTGGAACTGGCAGCTGTCATTGCTGGACCAGTGTGCTTCGTCTGCATCTCACTCATGTTGATGGTCTATATCTGCCACAACCGCACTGTCATTCACCATCGAGTGCCAAATGAAGAGGACCCTTCATTAGATCGCCCTTTTATTTCAGAGGGTACTACGTTGAAAGACTTAATTTATGATATGACAACGTCAGGTTCTGGCTCAGGTTTACCATTGCTTGTTCAGAGAACAATTGCGAGAACTATTGTGTTACAAGAAAGCATTGGCAAAGGTCGATTTGGAGAAGTTTGGAGAGGAAAGTGGCGGGGAGAAGAAGTTGCTGTTAAGATATTCTCCTCTAGAGAAGAACGTTCGTGGTTCCGTGAGGCAGAGATTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weitang Liao et al.
Frontiers in physiology, 11, 1093-1093 (2020-10-06)
Renal tubulointerstitial fibrosis is usually the final outcome of various end-stage renal diseases. Recent studies have reported that microRNAs (miRNAs) play an important role in renal fibrosis. However, the biological function of microRNAs in renal fibrosis is complicated and remains
Hilène Lin et al.
Scientific reports, 8(1), 10488-10488 (2018-07-12)
Cartilage loss in osteoarthritis (OA) results from altered local production of growth factors and metalloproteases (MMPs). Furin, an enzyme involved in the protein maturation of MMPs, might regulate chondrocyte function. Here, we tested the effect of furin on chondrocyte catabolism
Teppei Sugano et al.
British journal of cancer, 124(1), 228-236 (2020-11-28)
Metastasis is the primary cause of death in cancer patients, and its management is still a major challenge. Epithelial to mesenchymal transition (EMT) has been implicated in the process of cancer metastasis, and its pharmacological interference holds therapeutic promise. Traf2-
Lincan Duan et al.
PloS one, 13(7), e0200452-e0200452 (2018-07-12)
In the tumor progression, transforming growth factor β1 (TGFβ1) plays a critical role in tumorigenesis as well as metastasis. It is known that high plasma level of TGFβ1 in patients with advanced non-small cell lung cancer (NSCLC) is correlated with
Chao Li et al.
Communications biology, 3(1), 327-327 (2020-06-26)
Chronic inflammation plays a crucial role in vascular calcification. However, only a few studies have revealed the mechanisms underlying the development of inflammation under high-phosphate conditions in chronic kidney disease (CKD) patients. Here, we show that inflammation resulting from the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service