Skip to Content
Merck
All Photos(1)

Key Documents

D5763

Sigma-Aldrich

N-(2,4-Dinitroanilino)maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N4O6
CAS Number:
Molecular Weight:
278.18
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

storage temp.

−20°C

InChI

1S/C10H6N4O6/c15-9-3-4-10(16)12(9)11-7-2-1-6(13(17)18)5-8(7)14(19)20/h1-5,11H

InChI key

NBLHPQHLKDLIKS-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rao et al.
The Biochemical journal, 316 ( Pt 3), 771-775 (1996-06-15)
N-(2,4-Dinitroanilino)maleimide (DAM) reacts covalently with the thiol group of the xylanase from Chainia leading to complete inactivation in a manner similar to N-ethylmaleimide, but provides a reporter group at the active site of the enzyme. Increasing amounts of xylan offered
S S Keskar et al.
The Biochemical journal, 281 ( Pt 3), 601-605 (1992-02-01)
The kinetics of chemical modification of the xylanase from a thermotolerant Streptomyces T7 indicated the involvement of 1 mol of cysteine residue/mol of enzyme [Keskar, Srinivasan & Deshpande (1989) Biochem. J. 261, 49-55]. The chromophoric reagent N-(2,4-dinitroanilino)maleimide (DAM) reacts covalently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service