Skip to Content
Merck
All Photos(1)

Key Documents

D4546

Sigma-Aldrich

7-Deacetylforskolin 7-hemisuccinate

≥98%

Synonym(s):

7β-O-Hemisuccinyl-1α,6β,9α-trihydroxy-8,13-epoxy-labd-14-en-11-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O9
CAS Number:
Molecular Weight:
468.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

storage temp.

−20°C

SMILES string

[H][C@@]12[C@H](O)[C@H](OC(=O)CCC(O)=O)[C@@]3(C)O[C@](C)(CC(=O)[C@]3(O)[C@@]1(C)[C@@H](O)CCC2(C)C)C=C

Application

Water-soluble derivative of forskolin which may be coupled to agarose for the purification of adenylate cyclase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Pfeuffer et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(10), 3086-3090 (1985-05-01)
The guanosine 5'-[beta, gamma-imido]triphosphate (p[NH]ppG)-activated adenylate cyclase from rabbit myocardial membranes was purified approximately equal to 60,000-fold to a specific activity of 15 mumol X mg-1 X min-1 by Lubrol PX extraction, affinity chromatography, and gel permeation HPLC. The major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service