D2418
Denatonium saccharide
≥95% (perchloric acid titration)
About This Item
Assay
≥95% (perchloric acid titration)
SMILES string
O=C1[N-]S(=O)(=O)c2ccccc12.CC[N+](CC)(CC(=O)Nc3c(C)cccc3C)Cc4ccccc4
InChI
1S/C21H28N2O.C7H5NO3S/c1-5-23(6-2,15-19-13-8-7-9-14-19)16-20(24)22-21-17(3)11-10-12-18(21)4;9-7-5-3-1-2-4-6(5)12(10,11)8-7/h7-14H,5-6,15-16H2,1-4H3;1-4H,(H,8,9)
InChI key
AOVWCKBXCSVODH-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service