Skip to Content
Merck
All Photos(1)

Key Documents

D2013

Sigma-Aldrich

N,N-Dimethylbenzylamine

Synonym(s):

N-Benzyldimethylamine, BDMA, DMBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)2
CAS Number:
Molecular Weight:
135.21
Beilstein:
1099620
EC Number:
MDL number:
UNSPSC Code:
12352103

refractive index

n20/D 1.501 (lit.)

bp

183-184 °C/765 mmHg (lit.)

mp

−75 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CN(C)Cc1ccccc1

InChI

1S/C9H13N/c1-10(2)8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3

InChI key

XXBDWLFCJWSEKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Bard et al.
Journal of medicinal chemistry, 52(10), 3416-3419 (2009-04-29)
RPLC gains acceptance in pharmaceutical research for the rapid determination of lipophilicity but remains limited for the determination of partition coefficients of moderate to strong basic compounds under their neutral form because stationary phases are not compatible with high pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service