Skip to Content
Merck
All Photos(1)

Key Documents

CPS1300

Sigma-Aldrich

Chemiluminescent Peroxidase Substrate-1

sufficient for 300 mL substrate

Synonym(s):

CPS1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204

form

liquid

usage

sufficient for 300 mL substrate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Two component liquid peroxidase substrate system for Western blot detection. Picomole detection with superior signal-to-noise ratio. Component ratios can be adjusted for optimal signal duration, sensitivity and background.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhojit Paul et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 133, 111083-111083 (2021-01-01)
Apo-A1 is correlated with conditions like hyperlipidemia, cardiovascular diseases, high altitude pulmonary edema and etc. where hypoxia constitutes an important facet.Hypoxia causes oxidative stress, vaso-destructive and inflammatory outcomes.Apo-A1 is reported to have vasoprotective, anti-oxidative, anti-apoptotic, and anti-inflammatory effects. However, effects
Anamika Gangwar et al.
Scientific reports, 10(1), 7899-7899 (2020-05-15)
Intermittent hypoxia, initially associated with adverse effects of sleep apnea, has now metamorphosed into a module for improved sports performance. The regimen followed for improved sports performance is milder intermittent hypoxic training (IHT) as compared to chronic and severe intermittent
Babita Rahar et al.
High altitude medicine & biology, 20(1), 78-88 (2019-03-21)
High altitude exposure alters biochemical, metabolic, and physiological features of heart and skeletal muscles, and hence has pathological consequences in these tissues. Central to these hypoxia-associated biochemical/metabolic shuffling are energy deficit accumulation of free radicals and ensuing oxidative damage in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service