Skip to Content
Merck
All Photos(1)

Documents

CLS431175

Corning® Easy-Grip round, plastic, storage bottles

round clear polystyrene, sterile, bottle capacity 150 mL, cap (plug seal), case of 24

Synonym(s):

storage bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

cap (plug seal)
clear polystyrene bottle
cylindrical bottle
orange high-density polyethylene cap (plug seal)
round clear polystyrene

sterility

sterile
sterile

feature

cap
graduated
graduations
neck thread wide mouth

packaging

case of 24

manufacturer/tradename

Corning 431175

bottle capacity

150 mL

bottle neck diam.

~42 mm

neck diam.

45 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® Easy-Grip disposable storage bottles are ideal for storing media, buffers, and other aqueous solutions.
  • Features easy grip sides for convenient handling.
  • Plug seal caps (45 mm) provides an airtight seal and help minimize the risk of contamination.
  • Certified non pyrogenic.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service