Skip to Content
Merck
All Photos(1)

Documents

CLS3823

Corning® multicoated microplates

well size 96, black polystyrene, flat bottom wells clear, non-sterile, lid

Synonym(s):

3823, multicoated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13

material

black polystyrene
flat bottom wells clear
round wells

sterility

non-sterile

feature

lid
skirt

packaging

pack of 1 ea
case of 10 ea

manufacturer/tradename

Corning 3823

well size

96

well volume

360 μL

well working volume

75-200 μL

wells

96 wells

binding type

(multicoated (Collagen I, Fibronectin, Gelatin, Laminin, Poly-D-Lysine, TC-treated))

Looking for similar products? Visit Product Comparison Guide

General description

  • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescence and enabling readings down to 340nm
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Nonreversable lids with condensation rings to reduce contamination

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service