CLS354450
Corning® BioCoat™ Collagen I Culture Dish
diam. 100 mm
Synonym(s):
Cell Culture Dish, plastic cell culture dish, coated cell culture dish
Select a Size
£98.60
Select a Size
About This Item
£98.60
Recommended Products
feature
lid
packaging
pkg of 10 ea (1 pack )
manufacturer/tradename
Corning 354450
diam.
100 mm
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service