Skip to Content
Merck
All Photos(1)

Documents

CLS3535

Corning® Synthemax® II-SC substrate

vial of 10 mg

Synonym(s):

3535, II-SC substrate, synthemax, synthetic surface

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161501
NACRES:
NA.22

sterility

sterile; irradiated

packaging

vial of 10 mg

manufacturer/tradename

Corning 3535

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

The Corning Synthemax II-SC Substrate is an easy-to-use, self-coating material for creating a unique synthetic surface that mimics the natural cell environment. This surface supports the adhesion and expansion of stem cells in their undifferentiated state and directed differentiation into specialized cell types.

Legal Information

Corning is a registered trademark of Corning, Inc.
Synthemax is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Ritter et al.
Developmental cell, 50(5), 644-657 (2019-08-20)
Precisely controlled gene regulatory networks are required during embryonic development to give rise to various structures, including those of the cardiovascular system. Long non-coding RNA (lncRNA) loci are known to be important regulators of these genetic programs. We have identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service