Skip to Content
Merck
All Photos(1)

Key Documents

Lu2

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 99043003-DNA-5UG
pkg of vial of cells 99043003-1VL

ImmunoDon

BALB/c x C57 Bl/6

Myeloma

P3x63Ag8.653

Reactivity

Anti-DR11

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

The cell line Lu2 has been established by fusion of the myeloma P3x63Ag8.653 and spleen cells from BALB/c x C57Bl/6 female mice. Mice received a single i.p. injection with the B cell line LuMar expressing HLA A2.3/B18.5/C5/DR3,11.and B. pertussis three days before fusion. The cell line has been eradicated from mycoplasma using Cibrobay.

Immunogen

B cell line LuMar

Application

Immunofluorescence, cytotoxicity, immunoprecipitation

Culture Medium

RPMI 1640 + 2mM glutamine + 10% FBS

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service