Skip to Content
Merck
All Photos(1)

Documents

C2-Rev 7

94101907, rat liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

C2-Rev 7, 94101907

biological source

rat liver

growth mode

Adherent

karyotype

Hyperdiploid, 51 chromosomes

morphology

Epithelial

products

Albumin and hepatocyte markers

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat hepatoma [HGPRT-]

Cell Line Description

C2-Rev 7 is a revertant of the C2 variants, selected by virtue of its capacity to grow in glucose free medium (containing dialysed serum) where survival requires activity of the liver-specific gluconeogenic pathway. Expression of the entire set of liver specific functions is restored. Albumin and hepatocyte markers present.

Application

Gene expression studies.

Culture Medium

Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures (70%) 1:10 i.e. seeding at 9x1000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density is reached at 15x1,000,000 cells/10cm2 dish.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service