Skip to Content
Merck
All Photos(1)

Key Documents

VA-ES-BJ

94092303, human skin (bone marrow metastasis), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

VA-ES-BJ, 94092303

biological source

human skin (bone marrow metastasis)

growth mode

Adherent

karyotype

66-68 Aneuploidic, pentasomy #7, multiple trisomy

morphology

Epithelial

products

Granulocyte Macrophage Colony Stimulating Factor (GM-CSF)

receptors

Granulocyte Macrophage Colony Stimulating Factor (GM-CSF), AE, AE3, Fas, epithelial membrane antigen (EMA), Vimentin, P53

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human skin epithelioid sarcoma

Cell Line Description

Spontaneously transformed cells from a male patient with epithelioid sarcoma derived from bone marrow metastasis. Cells grow very rapidly and secrete GM-CSF. They have been shown to express AE-1 and AE-3 cytokeratins, epithelial membrane antigen, p53, human Fas and vimentin. VA-ES-BJ cells are sensitive to oxidising agents, busulfaxamine and protoporphyrin IX. Epithelial, spindle and giant cells.

Application

Study of tumours. Tumourigenic in nude mice.

Culture Medium

DMEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service