Key Documents
CV-1 Clone B5
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
UNSPSC Code:
41106514
biological source
African green monkey kidney
packaging
tube of 5 μg 90032601-DNA-5UG
pkg of vial of cells 90032601-1VL
growth mode
Adherent
karyotype
2n = 60, pseudodiploid
morphology
Fibroblast
receptors
Not specified
Cell Line Origin
Monkey African Green kidney fibroblast
Cell Line Description
The cells have been selected for sensitivity to SV40.
Culture Medium
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 2% Tricine (1M solution at pH7.4) + 10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:10, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)