Skip to Content
Merck
All Photos(1)

Key Documents

CMT 93

89111413, mouse colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CMT 93, 89111413

biological source

mouse colon

growth mode

Adherent

karyotype

Hyperdiploid,modal no. 50

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse rectum carcinoma

Cell Line Description

From a 19 month old male mouse (C57BL/1CRF) which had received an i.p. injection of MAMA each week for 18 months. 4th in vivo passage - source of explant culture. Cells form acini and junctional complexes and produce large tumours in nude mice (6 x 1,000,000 cells inoculum) after 1 month.

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.05% trypsin; 5% CO2; 37°C. Cells grow in clumps.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service