Skip to Content
Merck
All Photos(1)

Key Documents

AFRC IAH-CC38

Reactivity: Anti-CD6, bovine, 88022413

Sign Into View Organizational & Contract Pricing

Select a Size

88022413-DNA-5UG
£329.00
1 VIAL
£802.00

£329.00


Estimated to ship on02 April 2025Details



Select a Size

Change View
88022413-DNA-5UG
£329.00
1 VIAL
£802.00

About This Item

UNSPSC Code:
41106514

£329.00


Estimated to ship on02 April 2025Details


biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

NS1

Reactivity

Anti-CD6, bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against the BoCD6 antigen, which is the bovine equivalent of CD6.

Immunogen

Bovine thymocytes

Culture Medium

RPMI 1640 + 2mM glutamine + 1% Hypoxanthine, Thymidine (HT) supplement + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service