Skip to Content
Merck
All Photos(1)

Documents

RCC4plusVHL

3112703, human kidney, Epithelial

Synonym(s):

RCC4 VHL(+) Cells, RCC4-pVHL Cells, RCC4/VHL Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RCC4plusVHL, 03112703

biological source

human kidney

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Renal cell carcinoma cell line RCC4 stably transfected with pcDNA3-VHL

Cell Line Description

Renal cell carcinoma cell line RCC4 stably transfected with pcDNA3-VHL. The plasmid pcDNA3-VHL confers neomycin resistance and encodes the von Hippel-Lindau (VHL) tumour suppressor gene product pVHL. The original renal carcinoma cell line RCC4 is VHL-deficient. The RCC4 plus vector alone cell line (Sigma catalogue no. 03112702) can serve as a negative control cell line to study the effect of pVHL expression from pcDNA3-VHL. These cells are less tumourigenic than corresponding cell lines lacking pVHL function.

Application

Study of tumour suppressor gene product function - von Hippel-Lindau tumour suppressor gene. Cell responses to hypoxia.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 11,12
D16S539: 11
D5S818: 12
D7S820: 8.2
THO1: 9.3
TPOX: 11
vWA: 16

Culture Medium

DMEM + 10% FBS + 2 mM Glutamine + G418 0.5mg/ml

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 using 0.25% Trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service