Skip to Content
Merck
All Photos(1)

Key Documents

C8088

Sigma-Aldrich

Cantharidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O5
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

solid

solubility

DMSO: soluble
ethanol: soluble

SMILES string

OC(=O)[C@@H]1[C@H]2CC[C@H](O2)[C@@H]1C(O)=O

InChI

1S/C8H10O5/c9-7(10)5-3-1-2-4(13-3)6(5)8(11)12/h3-6H,1-2H2,(H,9,10)(H,11,12)/t3-,4+,5-,6+

InChI key

GXEKYRXVRROBEV-FBXFSONDSA-N

Biochem/physiol Actions

Useful for inhibiting PP1 and PP2A during purification of phosphorylated proteins, IC50 = 53 nM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y M Li et al.
Biochemical pharmacology, 46(8), 1435-1443 (1993-10-19)
The target site for cantharidin (CA) and its analogues was isolated recently from mouse liver and identified as protein phosphatase (PP2A) in the heterodimeric form known as PP2A2. The most toxic CA analogue, endothall thioanhydride (ETA) (mouse i.p. LD50 0.3
Koyuki Kondo et al.
PloS one, 10(12), e0144803-e0144803 (2015-12-23)
Upon desiccation, some tardigrades enter an ametabolic dehydrated state called anhydrobiosis and can survive a desiccated environment in this state. For successful transition to anhydrobiosis, some anhydrobiotic tardigrades require pre-incubation under high humidity conditions, a process called preconditioning, prior to
Y M Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(24), 11867-11870 (1992-12-15)
The toxic effects of cantharidin from blister beetles and its analogs, including the herbicide endothall, are attributable to their high affinity and specificity for a cantharidin-binding protein (CBP). An ammonium sulfate precipitate of mouse liver cytosol was purified by five
Masanori Ikeda et al.
Scientific reports, 7(1), 8794-8794 (2017-08-20)
For faithful chromosome segregation, the formation of stable kinetochore-microtubule attachment and its monitoring by the spindle assembly checkpoint (SAC) are coordinately regulated by mechanisms that are currently ill-defined. Here, we show that polo-like kinase 1 (Plk1), which is instrumental in
Hanne B Moeller et al.
Journal of cell science, 127(Pt 14), 3174-3183 (2014-05-31)
The post-translational modifications (PTMs) phosphorylation and ubiquitylation regulate plasma membrane protein function. Here, we examine the interplay between phosphorylation and ubiquitylation of the membrane protein aquaporin-2 (AQP2) and demonstrate that phosphorylation can override the previously suggested dominant endocytic signal of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service