Skip to Content
Merck
All Photos(2)

Key Documents

C6789

Sigma-Aldrich

Cholesteryl nervonate

~95%

Synonym(s):

3β-Hydroxy-5-cholestene 3-nervonate, 5-Cholesten-3β-ol 3-nervonate, Cholesteryl cis-15-tetracosenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H90O2
CAS Number:
Molecular Weight:
735.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

~95%

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCCCCCCCC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Sawzik et al.
Journal of lipid research, 25(8), 851-856 (1984-08-01)
At 123 K, cholesterol cis-15-tetracosenoate (cholesteryl nervonate, C51H90O2) is monoclinic, space group P2(1) with a = 12.948(5), b = 8.805(5), c = 42.98(5) A, beta = 105.93(3) degrees, [lambda(CuK alpha) = 1.5418 A], having two independent molecules (A) and (B)
Thomas J Millar et al.
Investigative ophthalmology & visual science, 53(8), 4710-4719 (2012-06-15)
The lipid layer of the tears has been studied in vivo using high resolution color microscopy (HRCM). The purpose of these experiments was to gain further insight into the structure of the lipid layer by applying HRCM to in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service