Skip to Content
Merck
All Photos(1)

Key Documents

C3705

Sigma-Aldrich

C Reactive Protein Fragment 174-185

≥97% (HPLC)

Synonym(s):

Ile-Tyr-Leu-Gly-Gly-Pro-Phe-Ser-Pro-Asn-Val-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H93N13O16
CAS Number:
Molecular Weight:
1276.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)NCC(=O)N2CCC[C@H]2C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CO)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C62H93N13O16/c1-9-36(8)51(64)59(87)70-42(28-38-19-21-39(77)22-20-38)54(82)67-40(25-33(2)3)53(81)66-30-49(79)65-31-50(80)74-23-13-17-46(74)57(85)68-41(27-37-15-11-10-12-16-37)55(83)72-45(32-76)61(89)75-24-14-18-47(75)58(86)69-43(29-48(63)78)56(84)73-52(35(6)7)60(88)71-44(62(90)91)26-34(4)5/h10-12,15-16,19-22,33-36,40-47,51-52,76-77H,9,13-14,17-18,23-32,64H2,1-8H3,(H2,63,78)(H,65,79)(H,66,81)(H,67,82)(H,68,85)(H,69,86)(H,70,87)(H,71,88)(H,72,83)(H,73,84)(H,90,91)/t36-,40-,41-,42-,43-,44-,45-,46-,47-,51-,52-/m0/s1

InChI key

KODGVWMUDZVVII-FVTMRLMCSA-N

Gene Information

human ... CRP(1401)

Amino Acid Sequence

Ile-Tyr-Leu-Gly-Gly-Pro-Phe-Ser-Pro-Asn-Val-Leu

Biochem/physiol Actions

C Reactive Protein (CRP) Fragment 174-185, CRP174-185, is a synthetic peptide fragment of CRP that induces the circulation of soluble IL-6 receptor (sIL-6R) and modulates chemotaxis. CRP174-185 and other CRP peptides such as 77-82 and 201-206 are use to resolve the function of amino acid sequences and domains withing CRP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Thomassen et al.
Journal of immunotherapy with emphasis on tumor immunology : official journal of the Society for Biological Therapy, 13(1), 1-6 (1993-01-01)
We have previously shown that native human C-reactive protein (CRP) produces antitumor effects in experimental animals, and that these effects are mediated primarily through macrophages. More recently, we have observed that RS-83277, a synthetic peptide derived from CRP, appears to
S A Jones et al.
The Journal of experimental medicine, 189(3), 599-604 (1999-02-02)
The soluble interleukin 6 receptor (sIL-6R) circulates at elevated levels in various diseases. This suggests that inflammatory mediators control sIL-6R release. Through examination of human neutrophils, it was found that the acute phase reactant C-reactive protein (CRP) activates a threefold
Stefan Rose-John et al.
Expert opinion on therapeutic targets, 11(5), 613-624 (2007-05-01)
IL-6 plays a pivotal role in immune responses and certain oncologic conditions. The intense investigation of its biological activity and function led to the discovery of two different IL-6-driven signalling pathways. Binding to the membrane-bound IL-6 receptor (mIL-6R, CD126) causes
Effect of human C-reactive protein on chemokine and chemotactic factor-induced neutrophil chemotaxis and signaling.
Zhong W, Zen Q, Tebo J, et al.
Journal of Immunology, 161, 2433-2540 (1998)
Tarek Khreiss et al.
The Journal of biological chemistry, 277(43), 40775-40781 (2002-08-29)
Human neutrophil granulocytes die rapidly, and their survival is contingent upon rescue from programmed cell death by signals from the environment. Here we report that a novel signal for delaying neutrophil apoptosis is the classic acute phase reactant, C-reactive protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service