Skip to Content
Merck
All Photos(1)

Key Documents

C106

Sigma-Aldrich

CGS-12066 maleate salt

solid

Synonym(s):

7-Trifluoromethyl-4-(4-methyl-1-piperazinyl)pyrrolo-[1,2-a]quinoxaline maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17F3N4 · C4H4O4
CAS Number:
Molecular Weight:
450.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: 1.4 mg/mL
ethanol: 1.8 mg/mL
0.1 M HCl: 4 mg/mL

SMILES string

OC(=O)\C=C/C(O)=O.CN1CCN(CC1)c2nc3cc(ccc3n4cccc24)C(F)(F)F

InChI

1S/C17H17F3N4.C4H4O4/c1-22-7-9-23(10-8-22)16-15-3-2-6-24(15)14-5-4-12(17(18,19)20)11-13(14)21-16;5-3(6)1-2-4(7)8/h2-6,11H,7-10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

ZBPAHEUAJMCLRD-BTJKTKAUSA-N

Gene Information

human ... HTR1B(3351)

Biochem/physiol Actions

5-HT1B serotonin receptor agonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Cioce et al.
Oncotarget, 5(12), 4129-4143 (2014-07-02)
Metabolic remodeling is a hallmark of cancer progression and may affect tumor chemoresistance. Here we investigated by 1H-NMR/PCA analysis the metabolic profile of chemoresistant breast cancer cell subpopulations (ALDHbright cells) and their response to metformin, a promising anticancer metabolic modulator.
Marianne Oksnes et al.
The Journal of clinical endocrinology and metabolism, 99(5), 1665-1674 (2014-02-13)
Conventional glucocorticoid replacement therapy fails to mimic the physiological cortisol rhythm, which may have implications for morbidity and mortality in patients with Addison's disease. The objective of the study was to compare the effects of continuous sc hydrocortisone infusion (CSHI)
Kenji Ohba et al.
PloS one, 9(5), e97787-e97787 (2014-05-27)
High prevalence of infection with high-risk human papilloma virus (HPV) ranging from 25 to 100% (average 31%) was observed in breast cancer (BC) patients in Singapore using novel DNA chip technology. Early stage of BC demonstrated higher HPV positivity, and
Mattia Bramini et al.
ACS nano, 8(5), 4304-4312 (2014-04-30)
Understanding nanoparticle interactions with the central nervous system, in particular the blood-brain barrier, is key to advances in therapeutics, as well as assessing the safety of nanoparticles. Challenges in achieving insights have been significant, even for relatively simple models. Here
Kriston Ganguli et al.
Pediatric research, 77(4), 528-535 (2015-01-13)
Bacterial contact in utero modulates fetal and neonatal immune responses. Maternal probiotic supplementation reduces the risk of immune-mediated disease in the infant. We investigated the immunomodulatory properties of live Lactobacillus rhamnosus GG and its SpaC pilus adhesin in human fetal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service