Skip to Content
Merck
All Photos(1)

Documents

BMSIPD4400B400F2

Sigma-Aldrich

Blender Bag With Full Filter

Synonym(s):

homogenization bag, homogenizer bag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.24

material

clear
low-density polyethylene

sterility

non-sterile

manufacturer/tradename

Benchmark Scientific IPD4400-B400F2

bag H × W

300 cm × 190 cm

capacity

400 mL

General description

Blender bag, 400ml, for use in BenchMasher 400 and other paddle blenders. Includes full nylon 70µm filter.

Application

Homogenization and blending of agricultural, food, pharmaceutical and laboratory samples.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service