Skip to Content
Merck
All Photos(1)

Key Documents

BM0031

Sigma-Aldrich

BMS-763534

≥98% (HPLC)

Synonym(s):

5-Chloro-1-[(1S)-1-cyclopropyl-2-methoxyethyl]-3-[[6-(difluoromethoxy)-2,5-dimethyl-3-pyridinyl]amino]-2(1H)-pyrazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21ClF2N4O3
CAS Number:
Molecular Weight:
414.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -40 to -55°, c = 0.8 in chloroform-d

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

SMILES string

O=C1C(NC2=CC(C)=C(OC(F)F)N=C2C)=NC(Cl)=CN1[C@H](COC)C3CC3

Biochem/physiol Actions

BMS-763534 is a pyrazinone-containing antagonist that targets corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1) with high affinity (IC50 = 0.26 & 0.4 nM against 150 pM ovine CRF for binding rat & human CRF-R1, respectively), potency (IC50 = 1.0 nM against 0.3 nM CRF-stimulated ATCH secretion from primary rat anterior pituitary cells), and selectivity, displaying little affinity toward porcine CRF-R2/CRF2 and 46 other receptor/channel/transporter proteins (IC50 >10 μM). BMS-763534 inhibits CRF-stimulated cAMP production in human Y-79 retinoblastoma cells in cultures (pA2 = 9.47) and exhibits anxiolytic efficacy in a rat situational anxiety model in vivo (0.5-3 mg/kg p.o.).
BMS-763534 is a pyrazinone-containing antagonist that targets corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1).

Features and Benefits

BMS-763534 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Lodge et al.
Neuropharmacology, 67, 284-293 (2012-11-24)
BMS-763534 is a potent (CRF(1) IC(50) = 0.4 nM) and selective (>1000-fold selectivity vs. all other sites tested) CRF(1) receptor antagonist (pA2 = 9.47 vs. CRF(1)-mediated cAMP production in Y79 cells). BMS-763534 accelerated the dissociation of (125)I-o-CRF from rat frontal cortex membrane CRF(1) receptors consistent
Richard A Hartz et al.
Bioorganic & medicinal chemistry letters, 20(6), 1890-1894 (2010-02-24)
A series of N(3)-pyridylpyrazinones was investigated as corticotropin-releasing factor-1 receptor antagonists. It was observed that the binding affinity of analogues containing a pyridyl group was influenced not only by the substitution pattern on the pyridyl group, but also by the
Richard A Hartz et al.
Journal of medicinal chemistry, 52(23), 7653-7668 (2009-12-04)
Detailed metabolic characterization of 8, an earlier lead pyrazinone-based corticotropin-releasing factor-1 (CRF(1)) receptor antagonist, revealed that this compound formed significant levels of reactive metabolites, as measured by in vivo and in vitro biotransformation studies. This was of particular concern due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service