Skip to Content
Merck
All Photos(1)

Documents

B6272

Sigma-Aldrich

8-Bromoadenosine

powder

Synonym(s):

6-Amino-8-bromopurine riboside, 8-Bromoadenine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12BrN5O4
CAS Number:
Molecular Weight:
346.14
Beilstein:
627737
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥98% (HPLC)

Quality Level

form

powder

optical activity

[α]/D -55 to -50°, c = 22-28 mg/mL in DMSO

color

white

mp

210-212 °C (dec.) (lit.)

solubility

1 M HCl: 49-51 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Nc1ncnc2n([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c(Br)nc12

InChI

1S/C10H12BrN5O4/c11-10-15-4-7(12)13-2-14-8(4)16(10)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,13,14)/t3-,5-,6-,9-/m1/s1

InChI key

VJUPMOPLUQHMLE-UUOKFMHZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose P Carrascosa et al.
Reproductive sciences (Thousand Oaks, Calif.), 25(7), 985-999 (2017-10-07)
Intrauterine devices (IUDs) have been widely used to prevent pregnancies with great efficacy during decades. It has been demonstrated that IUD alters the endometrial gene expression, but there is no scientific data about how copper, a metal commonly used in
Ruban Rex Peter Durairaj et al.
Molecular human reproduction, 23(7), 478-487 (2017-04-13)
Is implantation failure following ART associated with a perturbed decidual response in endometrial stromal cells (EnSCs)? Dynamic changes in the secretome of decidualizing EnSCs underpin the transition of a hostile to a supportive endometrial microenvironment for embryo implantation; perturbation in
Clara Lazzaretti et al.
Reproductive biomedicine online, 38(5), 816-824 (2019-03-27)
What is the cumulative effect of two follicle-stimulating hormone receptor (FSHR) mutations in spontaneous ovarian hyperstimulation syndrome (sOHSS) pathogenesis? Are these mutations in the mono- or biallelic state? Two FSHR mutations were found in a pregnant patient affected by sOHSS
Monir Hossain et al.
BMB reports, 53(3), 142-147 (2019-08-14)
Lipid accumulation in white adipose tissue is the key contributor to the obesity and orchestrates numerous metabolic health problems such as type 2 diabetes, hypertension, atherosclerosis, and cancer. Nonetheless, the prevention and treatment of obesity are still inadequate. Recently, scientists
Chryssostomos Chatgilialoglu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(5), 1249-1255 (2004-03-10)
The reaction of hydrated electrons (e(-)(aq) with 8-bromoadenosine 7 has been investigated by radiolytic methods coupled with product studies. Pulse radiolysis revealed that one-electron reductive cleavage of the C-Br bond gives the C8 radical 8 followed by a fast radical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service