Skip to Content
Merck
All Photos(1)

Key Documents

B2397

Sigma-Aldrich

Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside

Synonym(s):

α-D-GlcNAc-1→OCH2Ph, Benzyl N-acetyl-α-D-glucosaminide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO6
CAS Number:
Molecular Weight:
311.33
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OCc2ccccc2

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service