Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT200O

Screw cap tubes and caps with O-rings

size 2.0 mL, orange, pkg of 8x500caps/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc SCT-200-O
Corning

packaging

pkg of 8x500caps/cs

size

2.0 mL

color

orange

Looking for similar products? Visit Product Comparison Guide

General description

Product is only available in China.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service