Skip to Content
Merck
All Photos(2)

Key Documents

AV53702

Sigma-Aldrich

Anti-FEM1B (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp451E0710, Anti-FIAA, Anti-Fem-1 homolog b (C. elegans)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£328.00

£328.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
£328.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£328.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

bovine, dog, horse, guinea pig, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FEM1B(10116)

Immunogen

Synthetic peptide directed towards the middle region of human FEM1B

Application

Anti-FEM1B (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

FEM1B [fem-1 homolog b (C. elegans)] gene encodes a protein which is a homolog of feminization-1 (FEM-1), a protein involved in the sex-determination pathway of the nematode Caenorhabditis elegans. It stimulates ubiquitylation of Gli1 as well as inhibits its transcriptional activity.[1] It is a proapoptotic protein that facilitates proteasome inhibitor-induced apoptosis of human colon cancer cells.[2] Additionally, FEM1B serves as an adapter or mediator protein that links CHK1 and Rad9 and facilitates checkpoint signaling induced by replication stress.[3]

Sequence

Synthetic peptide located within the following region: FQDGDNILEKEVLPPIHAYGNRTECRNPQELESIRQDRDALHMEGLIVRE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew S Gilder et al.
Biochemical and biophysical research communications, 440(3), 431-436 (2013-10-01)
The mammalian Fem1b gene encodes a homolog of FEM-1, a protein in the sex-determination pathway of the nematode Caenorhabditis elegans. Fem1b and FEM-1 proteins each contain a VHL-box motif that mediates their interaction with certain E3 ubiquitin ligase complexes. In
M Cecilia Subauste et al.
Molecular carcinogenesis, 49(2), 105-113 (2009-11-13)
In the treatment of colon cancer, the development of resistance to apoptosis is a major factor in resistance to therapy. New molecular approaches to overcome apoptosis resistance, such as selectively upregulating proapoptotic proteins, are needed in colon cancer therapy. In
T-P Sun et al.
Oncogene, 28(18), 1971-1981 (2009-03-31)
Human checkpoint kinase 1 (CHK1) is an essential kinase required to preserve genome stability, and is activated by DNA replication blockage through the ataxia-telangiectasia-mutated-and-Rad3-related (ATR)/ATRIP-signaling pathway. In this report, we show that a novel CHK1-interacting protein, FEM1B (human homologue of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service