Skip to Content
Merck
All Photos(2)

Key Documents

AV49163

Sigma-Aldrich

Anti-RABGGTA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Rab geranylgeranyltransferase, α subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£437.00

£437.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
£437.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£437.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65 kDa

species reactivity

human, rat, guinea pig, dog, bovine, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RABGGTA(5875)

Immunogen

Synthetic peptide directed towards the N terminal region of human RABGGTA

Application

Anti-RABGGTA antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

Rab geranylgeranyltransferase, alpha subunit (RABGGTA; PTAR3) an enzyme that attaches geranylgeranyl groups to Rab proteins. The activity of this enzyme is essential for hemostasis and platelet synthesis. Polymorphisms in RABGGTA gene have been observed in patients with Hermansky-Pudlak syndrome.

Sequence

Synthetic peptide located within the following region: ELAFTDSLITRNFSNYSSWHYRSCLLPQLHPQPDSGPQGRLPEDVLLKEL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Li et al.
Molecular genetics and metabolism, 71(4), 599-608 (2001-01-04)
Hermansky-Pudlak syndrome (HPS) is a recessively inherited disease with dysfunction of several related subcellular organelles including platelet-dense granules, melanosomes, and lysosomes. Our recent identification of the mutation in murine Rab geranylgeranyl transferase alpha-subunit gene (Rabggta) in one mouse model of
J C Detter et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(8), 4144-4149 (2000-03-29)
Few molecular events important to platelet biogenesis have been identified. Mice homozygous for the spontaneous, recessive mutation gunmetal (gm) have prolonged bleeding, thrombocytopenia, and reduced platelet alpha- and delta-granule contents. Here we show by positional cloning that gm results from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service