Skip to Content
Merck
All Photos(4)

Key Documents

AV41733

Sigma-Aldrich

Anti-KRT17 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-K17, Anti-Keratin 17, Anti-PC, Anti-PC2, Anti-PCHC1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£437.00

£437.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
£437.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£437.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

dog, rabbit, mouse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT17(3872)

Immunogen

Synthetic peptide directed towards the C terminal region of human KRT17

Biochem/physiol Actions

KRT17 is type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in its gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.KRT17 encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.

Sequence

Synthetic peptide located within the following region: IATYRRLLEGEDAHLTQYKKEPVTTRQVRTIVEEVQDGKVISSREQVHQT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service