Skip to Content
Merck
All Photos(1)

Key Documents

AC-0356A

Sigma-Aldrich

MAGEC2 (CT10) (EP405) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
£263.00

£263.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.1 ML
£263.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£263.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

The Melanoma-associated antigen C2, MAGEC2, also known as CT10, was identified as a novel Cancer/testis (CT)-antigen originally identified in the SK-MEL-37 melanoma cell line. MAGEC2 is a member of the MAGEC gene family, mapping to chromosome Xq26-q27 and is closely related to MAGEC1 (CT7). CT-X antigens are thought to repress the expression of certain genes necessary for cellular differentiation. MAGEC2 expression is restricted to the testis in normal tissues. Male germ cells highly express MAGEC2 during early phases of spermatogenesis. Immature spermatogonia and spermatocyte demonstrated strong MAGEC2 nuclear expression and is decreased in spermatids. MAGEC2 is not expressed in mature sperm and surrounding stromal, Sertoli, and Leydig cells. Bode, et al. examined 325 testicular germ cell tumors and found MAGEC2 in most seminoma (94%), spermatocytic seminoma (100%) and intratubular germ cell neoplasia (IGCNU) (100%). Conversely, MAGEC2 expression were uncommon in embryonal carcinoma, yolk sac tumor, teratoma and choriocarcinoma. MAGEC2 protein expression is a sensitive marker for seminomas, spermatocytic seminomas, and IGCNU. Use of this marker may be helpful in the diagnosing seminoma (positive) from embryonal carcinoma (negative) when assessed in a panel of germ cell tumor markers including OCT4, KIT, PLAP, CD30, SOX2 and SOX17. Additionally, MAGEC2 was identified as a prognostic marker in a variety of tumor types. MAGEC2 positive tumors were frequently associated with reduced recurrence free and overall survival rates in prostate, hepatocellular, triple-negative breast, and non-small cell lung carcinomas.

Application

Immunohistochemistry (formalin-fixed, paraffin-embedded sections)

Quality


IVD

IVD

IVD

RUO

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service