Skip to Content
Merck
All Photos(1)

Key Documents

A2783

Sigma-Aldrich

4-(4-Dimethylaminophenylazo)aniline

Synonym(s):

4-Amino-4′-dimethylaminoazobenzene, N,N-Dimethyl-4,4′-azodianiline, p,p′-DMPA-aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N4
CAS Number:
Molecular Weight:
240.30
Beilstein:
748253
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

190-195 °C (dec.)

storage temp.

2-8°C

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccc(N)cc2

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service