Skip to Content
Merck
All Photos(1)

Key Documents

A2267

Sigma-Aldrich

1-Amino-1-deoxy-β-D-galactose

Synonym(s):

β-D-Galactosylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5
CAS Number:
Molecular Weight:
179.17
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥98% (TLC)

Quality Level

form

powder

optical activity

[α]/D 60.00 to 64.00°, c = 18.00-22.00 mg/mL in water

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-8°C

SMILES string

NC1OC(CO)C(O)C(O)C1O

InChI

1S/C6H13NO5/c7-6-5(11)4(10)3(9)2(1-8)12-6/h2-6,8-11H,1,7H2

InChI key

WCWOEQFAYSXBRK-UHFFFAOYSA-N

Application

β-D-Galactosylamine, a galactose analogue, is used as a competitive inhibitor to help isolate, purify, identify, differentiate and characterize β-D-galactosidase(s) and galactose oxidase(s).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

R A DiCioccio et al.
Carbohydrate research, 127(1), 109-120 (1984-04-02)
A beta-D-galactosidase from bovine liver was purified to apparent homogeneity. The major purification step was affinity chromatography on a column of D-galactose attached to a Sepharose support activated with divinyl sulfone. Affinity media prepared by binding ligands to Sepharose activated
N Monji et al.
Research communications in chemical pathology and pharmacology, 26(1), 187-196 (1979-10-01)
We have developed a new method for separation of antibody bound and unbound enzyme conjugates. The technique as applied to the assay of choriomammotropin involves the use of beta-D-galactosylamine bound to agarose to separate the unbound choriomammotropin-beta-galactosidase conjugates for antibody
C F Mazitsos et al.
Journal of chromatography. A, 954(1-2), 137-150 (2002-06-13)
Two anthraquinone galactosyl-biomimetic dye-ligands comprising, as terminal biomimetic moiety, galactose analogues (1-amino-1-deoxy-beta-D-galactose and D(+)-galactosamine) were designed for the enzyme galactose oxidase (GAO), using molecular modelling, synthesized and characterized. The biomimetic ligands were immobilized on agarose beads and the affinity adsorbents
Bo-Yu Li et al.
Carbohydrate research, 345(12), 1708-1712 (2010-06-30)
The highly diastereoselective InCl(3)-catalyzed aza-Friedel-Crafts reaction of substituted indoles with aldimines generated from Kunz's amine was studied. The reaction afforded the desired product in good to high yields with up to >19:1 diastereoselective ratios. The O-pivaloylated beta-D-galactosyl moiety could not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service