Skip to Content
Merck
All Photos(1)

Key Documents

A172

Sigma-Aldrich

1-Aminocyclobutane-cis-1,3-dicarboxylic acid

Synonym(s):

1-Amino-cis-1,3-cyclobutanedicarboxylic acid, cis-ACDA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

solubility

0.1 M NaOH: 12 mg/mL
0.1 M HCl: 14 mg/mL
H2O: 5.2 mg/mL
methanol: insoluble

Biochem/physiol Actions

Potent and selective NMDA receptor agonist

Other Notes

Note: Under IUPAC nomenclature, this compound is designated as trans

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D Allan et al.
Journal of medicinal chemistry, 33(10), 2905-2915 (1990-10-01)
We report the synthesis of a series of 3-carboxy-, 3-(carboxymethyl)-, 3-(omega-phosphonoalkyl)-1-aminocyclobutane-1-carboxylic acids for evaluation as agonists or antagonists of neurotransmission at excitatory amino acid receptors, particularly N-methyl-D-aspartic acid (NMDA) receptors. The compounds were evaluated as agonists on their ability to
Barbara Banelli et al.
Cell cycle (Georgetown, Tex.), 14(21), 3418-3429 (2015-11-15)
Notwithstanding current multimodal treatment, including surgery, radiotherapy and chemotherapy with temozolomide (TMZ), median survival of glioblastoma (GBM) patients is about 14 months, due to the rapid emergence of cell clones resistant to treatment. Therefore, understanding the mechanisms underlying chemoresistance is
Xiujuan Shi et al.
Molecular medicine reports, 11(5), 3533-3538 (2015-01-13)
Spindle and kinetochore‑associated protein 1 (SKA1) is an important component of the human kinetochore, which plays a key role in mitosis. The resent study was designed to investigate the role of SKA1 in human glioblastoma. The results of the present
Yuri Chikano et al.
Molecular cancer therapeutics, 14(2), 564-574 (2014-12-17)
The failure of current treatment options for glioblastoma stems from their inability to control tumor cell proliferation and invasion. Biologically targeted therapies offer great hope and one promising target is glycogen synthase kinase-3β (GSK3β), implicated in various diseases, including cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service