Skip to Content
Merck
All Photos(1)

Documents

A158

Supelco

Arcaine sulfate salt

Synonym(s):

1,4-Diguanidinobutane sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16N6 · H2O4S
CAS Number:
Molecular Weight:
270.31
MDL number:
UNSPSC Code:
41116107

SMILES string

OS(O)(=O)=O.NC(=N)NCCCCNC(N)=N

InChI

1S/C6H16N6.H2O4S/c7-5(8)11-3-1-2-4-12-6(9)10;1-5(2,3)4/h1-4H2,(H4,7,8,11)(H4,9,10,12);(H2,1,2,3,4)

InChI key

RWTGFMPOODRXIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service