Skip to Content
Merck
All Photos(1)

Documents

A120

Sigma-Aldrich

5-Aminovaleric acid hydrochloride

solid

Synonym(s):

5-Aminopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

form

solid

color

white

solubility

H2O: soluble
methanol: soluble

Biochem/physiol Actions

Weak GABAB receptor antagonist.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Schwarz et al.
Experimental brain research, 70(3), 618-626 (1988-01-01)
The action of delta-aminovaleric acid (AVA) on the muscle relaxant properties of baclofen, a GABAB receptor agonist, was investigated in two experimental models: (1) the pathologically increased muscle tone of the gastrocnemius muscle in spastic mutant Han-Wistar rats and (2)
Pharmacological specificity of synaptosomal and synaptic membrane gamma-aminobutyric acid (GABA) transport processes.
S L Early et al.
Biochemical pharmacology, 30(10), 1105-1113 (1981-05-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service