Skip to Content
Merck
All Photos(1)

Key Documents

A0483

Sigma-Aldrich

Monoclonal Anti-Actin (20-33) antibody produced in rabbit

clone SIG2-AC2, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SIG2-AC2, monoclonal

mol wt

antigen ~42 kDa

species reactivity

chicken, rat, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1:400
western blot: 1:50-1:100 using chicken gizzard total extract

isotype

IgG

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)

General description

Monoclonal Anti-Actin (20-33) (rabbit IgG) is a rabbit monoclonal derived from the hybridoma SIG2-AC2 produced by the fusion of rabbit myeloma cells and splenocytes from rabbits immunized with a synthetic peptide. Actin is one of the most conserved eukaryotic proteins and is expressed in mammals and birds with at least six isoforms characterized by electrophoresis and amino acid sequence analysis. There are three α-actins (skeletal, cardiac and smooth muscle), one β-actin (β-non-muscle) and two γ-actins (γ-smooth muscle and g-non-muscle).

Immunogen

synthetic peptide corresponding to amino acids 20-33 of actin, attached to a multiple antigen peptide (MAP) backbone.

Application

Monoclonal Anti-Actin (20-33) antibody produced in rabbit has been used in:
  • western blotting[1]
  • enzyme-linked immunosorbent assay (ELISA)
  • immunohistochemistry
  • immunocytochemistry

Physical form

Supplied as ascites fluid containing 15 mM sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of MCPH1 inhibits uncontrolled cell growth by promoting cell apoptosis and arresting the cell cycle in S and G2/M phase in lung cancer cells
Zhou L, et al.
Oncology Letters, 11(1), 365-372 (2016)
PP2A blockade inhibits autophagy and causes intraneuronal accumulation of ubiquitinated proteins
Magnaudeix A, et al.
Neurobiology of Aging, 34(3), 770-790 (2013)
The Potential Roles of Actin in The Nucleus
Falahzadeh K, et al.
Cell Journal, 17(1), 7-7 (2015)
Melania Cruciani et al.
Frontiers in immunology, 10, 2622-2622 (2019-11-30)
Understanding Staphylococcus aureus (S. aureus)-host immune system interaction is crucial to meet the tremendous medical need associated with this life-threatening bacterial infection. Given the crucial role of dendritic cells (DC) in dictating immune responses upon microbial challenge, we investigated how
Esther Klaile et al.
Cell cycle (Georgetown, Tex.), 7(20), 3180-3186 (2008-10-10)
In order to maintain genomic integrity during mitosis, cells assemble the mitotic spindle to separate sister chromosomes to the two daughter cells. A variety of motor- and non motor-proteins are involved in the organization and regulation of this complex apparatus.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service