74089
1-Amino-1-deoxy-scyllo-inositol
≥95.0% (TLC)
Synonym(s):
scyllo-Inosamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13NO5
CAS Number:
Molecular Weight:
179.17
Beilstein:
2802457
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
Quality Level
Assay
≥95.0% (TLC)
form
powder
storage temp.
2-8°C
SMILES string
O[C@@H]1[C@@H](N)[C@H](O)[C@@H](O)[C@H](O)[C@H]1O
InChI
1S/C6H13NO5/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6,8-12H,7H2/t1-,2-,3+,4+,5-,6-
InChI key
JXAOTICXQLILTC-CDRYSYESSA-N
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Stereochemical recognition of doubly functional aminotransferase in 2-deoxystreptamine biosynthesis.
Kenichi Yokoyama et al.
Journal of the American Chemical Society, 127(16), 5869-5874 (2005-04-21)
The doubly functional aminotransferase BtrS in the 2-deoxystreptamine (DOS) biosynthesis, in which two transaminations are involved, was characterized by a genetic as well as a chemical approach with the heterologously expressed enzyme. The gene disruption study clearly showed that BtrS
Lucía Díaz et al.
Journal of medicinal chemistry, 54(7), 2069-2079 (2011-03-05)
A library of aminocyclitols derived from CuAAC reaction between N-propargylaminocyclitol 4 and a series of azides [1-25] is described and tested against GCase. Azides have been chosen from a large collection of potential candidates that has been filtered according to
C P Saint et al.
Journal of bacteriology, 175(16), 5205-5215 (1993-08-01)
Rhizopines are selective growth substrates synthesized in nodules only by strains of rhizobia capable of their catabolism. We report the isolation and study of genes for the synthesis and catabolism of a new rhizopine, scyllo-inosamine (sIa), from alfalfa nodules induced
Christopher P Phenix et al.
Chembiochem : a European journal of chemical biology, 9(10), 1591-1602 (2008-06-07)
MosA is an enzyme from Sinorhizobium meliloti L5-30, a beneficial soil bacterium that forms a symbiotic relationship with leguminous plants. MosA was proposed to catalyze the conversion of scyllo-inosamine to 3-O-methyl-scyllo-inosamine (compounds known as rhizopines), despite the MosA sequence showing
Meritxell Egido-Gabás et al.
Organic & biomolecular chemistry, 3(7), 1195-1201 (2005-03-24)
A series of 13 aminocyclitol derivatives belonging to two different families is described. Their configuration is governed by the regio- and stereocontrolled epoxide opening of a suitably protected conduritol-B epoxide. Studies on several glycosyl processing enzymes indicate that some of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service