Skip to Content
Merck
All Photos(1)

Documents

60127

Sigma-Aldrich

Potassium chlorate

purum, ≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

form

solid

mp

356 °C (lit.)

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Khan et al.
Biochimica et biophysica acta, 1481(2), 229-236 (2000-10-06)
Characterization of conformational transition and folding intermediates is central to the study of protein folding. We studied the effect of various alcohols (trifluoroethanol (TFE), butanol, propanol, ethanol and methanol) and salts (K(3)FeCN(6), Na(2)SO(4), KClO(4) and KCl) on the acid-induced state
Vicente Parra et al.
Physical chemistry chemical physics : PCCP, 9(18), 2266-2273 (2007-05-10)
Here we report on the electroactivity properties of Langmuir-Blodgett (LB) films of the hole-transport molecule 4,4',4''-tris[3-methylphenyl(phenyl)amino] triphenylamine (m-MTDATA). Fairly stable Langmuir films at the air-water interface are accomplished, despite the non-amphiphilic character of the molecule. The reflection-absorption infrared spectroscopy (RAIRS)
John J Bosnjak et al.
American journal of physiology. Heart and circulatory physiology, 284(4), H1080-H1086 (2003-02-22)
Thrombin (Thromb), activated as part of the clotting cascade, dilates conduit arteries through an endothelial pertussis toxin (PTX)-sensitive G-protein receptor and releases nitric oxide (NO). Thromb also acts on downstream microvessels. Therefore, we examined whether Thromb dilates human coronary arterioles
Kazutomo Ishi et al.
Bioscience, biotechnology, and biochemistry, 69(12), 2463-2465 (2005-12-27)
The nitrate reductase gene (niaD) is the most frequently utilized as a selectable marker for homologous integration at the niaD locus of Aspergillus oryzae. In this study we developed a method for curing of the niaD-based plasmid integrated on the
Paola González Audino et al.
Pest management science, 58(2), 183-189 (2002-02-21)
The stability to heart of cis-permethrin and beta-cypermethrin in the solid phase was studied and the decomposition products identified. Samples heated at 210 degrees C in an oven in the dark showed that, in the absence of potassium chlorate (the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service