Skip to Content
Merck
All Photos(1)

Key Documents

52283

Sigma-Aldrich

Anti-Mouse IgG−Abberior® STAR RED antibody produced in goat

for STED application

Synonym(s):

Abberior® STAR RED-Anti-Mouse-IgG antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.45

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

~1 mg/mL

fluorescence

λex 638 nm; λem 655 nm in PBS, pH 7.4

storage temp.

−20°C

General description

Abberior STAR RED is a well-known red dye, named also KK114 in literature. It was developed for STED and confocal imaging in the red spectral region. It shows an exceptional brightness and low background, delivering stunning STED images. The dye works extremely well with Abberior Instruments STEDYCON and Expert Line microscopes and Leica STED microscopes. Abberior STAR RED can substitute dyes like Atto 647N, Alexa Fluor 647, or Cy®5. It can be excited with lasers at wavelength between 630 - 650 nm. For STED, a depletion wavelength around 750 - 800 nm is recommended. Abberior STAR RED is the ideal partner for Abberior STAR 580 to obtain optimal 2 Color STED results. Best results are obtained with freshly prepared samples.


Photophysical properties (carboxylic acid):
Absorption Maximum, λex [nm]: 638 (PBS pH 7.4), 638 (H2O), 634 (MeOH)
Extinction Coefficient, εmax [M-1cm-1]: 120 000 (PBS pH 7.4), 125 000 (H2O), 115 000 (MeOH)
Correction Factor, CF260 = ε260max: 0,16 (PBS pH 7.4)
Correction Factor, CF280 = ε280max: 0,32 (PBS pH 7.4)
Fluorescence Maximum, λem [nm]: 655 (PBS pH 7.4), 655 (H2O), 654 (MeOH)
Recommended STED Wavelength, λ [nm]: 750-800
Fluorescence Quantum Yield, λ: 0,90 (PBS pH 7.4)
Fluorescence Lifetime, τ [ns]: 3,4 (PBS pH 7.4)

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

May contain significant amounts of BSA as stabilizer
unconjugated dye ≤5% of total fluorescence

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Atto is a trademark of Atto-Tec GmbH
Cy is a registered trademark of Cytiva
abberior is a registered trademark of Abberior GmbH

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinem K Saka et al.
Nature communications, 5, 4509-4509 (2014-07-26)
Most proteins have uneven distributions in the plasma membrane. Broadly speaking, this may be caused by mechanisms specific to each protein, or may be a consequence of a general pattern that affects the distribution of all membrane proteins. The latter
Eulashini Chuntharpursat-Bon et al.
Communications biology, 6(1), 358-358 (2023-04-04)
Two prominent concepts for the sensing of shear stress by endothelium are the PIEZO1 channel as a mediator of mechanically activated calcium ion entry and the PECAM1 cell adhesion molecule as the apex of a triad with CDH5 and VGFR2.
Kirill Kolmakov et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(1), 146-157 (2013-12-18)
The synthesis, reactivity, and photophysical properties of new rhodamines with intense red fluorescence, two polar residues (hydroxyls, primary phosphates, or sulfonic acid groups), and improved hydrolytic stability of the amino-reactive sites (NHS esters or mixed N-succinimidyl carbonates) are reported. All
Alf Honigmann et al.
eLife, 3, e01671-e01671 (2014-03-20)
The eukaryotic cell membrane is connected to a dense actin rich cortex. We present FCS and STED experiments showing that dense membrane bound actin networks have severe influence on lipid phase separation. A minimal actin cortex was bound to a
Maksim V Sednev et al.
Bioconjugate chemistry, 24(4), 690-700 (2013-03-23)
Asymmetric hybrid fluorophores are built from the structural elements of two (or even more) symmetric dyes and can develop valuable new features which their parents do not possess. A new hybrid carborhodol dye was obtained by the combination of fluorescein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service