Skip to Content
Merck
All Photos(1)

Key Documents

43405

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

≥99.0% (HPLC), from yeast

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
39200202

biological source

yeast

Assay

≥99.0% (HPLC)

impurities

≤0.1% solvent
≤10% water

solubility

H2O: 50 mg/mL, clear, colorless

absorbance ratio

A280/260 nm 0.22 ±0.02
A250/260 nm 0.83 ±0.03

suitability

in accordance for determination of ADH

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nicotinamide adenine dinucleotide (NAD) and NADH form a redox pair. NAD/NADH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications. NAD is also involved in post-translational (Poly(ADP-ribose) polymerization) modifications of proteins.

Biochem/physiol Actions

Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mi Young Kim et al.
Genes & development, 19(17), 1951-1967 (2005-09-06)
Poly(ADP-ribose) (PAR) and the PAR polymerases (PARPs) that catalyze its synthesis from donor nicotinamide adenine dinucleotide (NAD+) molecules have received considerable attention in the recent literature. Poly(ADP-ribosyl)ation (PARylation) plays diverse roles in many molecular and cellular processes, including DNA damage
D D'Amours et al.
The Biochemical journal, 342 ( Pt 2), 249-268 (1999-08-24)
Poly(ADP-ribosyl)ation is a post-translational modification of proteins. During this process, molecules of ADP-ribose are added successively on to acceptor proteins to form branched polymers. This modification is transient but very extensive in vivo, as polymer chains can reach more than
Ana P Gomes et al.
Cell, 155(7), 1624-1638 (2013-12-24)
Ever since eukaryotes subsumed the bacterial ancestor of mitochondria, the nuclear and mitochondrial genomes have had to closely coordinate their activities, as each encode different subunits of the oxidative phosphorylation (OXPHOS) system. Mitochondrial dysfunction is a hallmark of aging, but
Teigo Asai et al.
Organic letters, 15(8), 2058-2061 (2013-04-13)
Graphiopsis chlorocephala was separated from the surface-sterilized healthy leaves of Paeonia lactiflora (Paeoniaceae) and cultivated with nicotinamide (an NAD(+)-dependent HDAC inhibitor). The culture conditions significantly enhanced secondary metabolite production in the fungus and led to the isolation of a structurally
Jeerus Sucharitakul et al.
The Journal of biological chemistry, 288(49), 35210-35221 (2013-10-17)
3-Hydroxybenzoate 6-hydroxylase (3HB6H) from Rhodococcus jostii RHA1 is an NADH-specific flavoprotein monooxygenase that catalyzes the para-hydroxylation of 3-hydroxybenzoate (3HB) to form 2,5-dihydroxybenzoate (2,5-DHB). Based on results from stopped-flow spectrophotometry, the reduced enzyme-3HB complex reacts with oxygen to form a C4a-peroxy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service