Skip to Content
Merck
All Photos(1)

Key Documents

410R-1

Sigma-Aldrich

Cytokeratin 10 (EP97) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP97, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (410R-14)
vial of 0.5 mL concentrate (410R-15)
bottle of 1.0 mL predilute (410R-17)
vial of 1.0 mL concentrate (410R-16)
bottle of 7.0 mL predilute (410R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

squamous cell carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... KRT10(3858)

General description

Cytokeratin 10 (CK10) belongs to type I and acidic keratin family.[1] It is low molecular weight keratin that is usually expressed in suprabasal keratinocytes and can also be seen in suprabasal cells of internal noncornifying, stratified squamous epithelia.[1]In addition, CK10 is reported to be a typical component of cells of eccrine sweat gland ducts and sebaceous cells.[2]Anti-CK10 is helpful in the identification of well differentiated squamous cell carcinoma derived from skin or from internal organs.[2][3]However, CK10 was found to be sparse in poorly differentiated squamous cell carcinoma, even though it has been reported that focal CK10 expression has been found in 50% of cases of oral and pharyngeal squamous cell carcinoma.[3]Anti-CK10 has demonstrated utility in the differentiation of extra-mammary Paget′s disease (negative) from basal cell carcinoma and squamous cell carcinoma (positive).[3]

Quality


IVD

IVD

IVD

RUO

Linkage

Cytokeratin 10 Positive Control Slides, Product No. 410S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Moll et al.
Histochemistry and cell biology, 129(6), 705-733 (2008-05-08)
The keratins are the typical intermediate filament proteins of epithelia, showing an outstanding degree of molecular diversity. Heteropolymeric filaments are formed by pairing of type I and type II molecules. In humans 54 functional keratin genes exist. They are expressed
Cytokeratins as markers of differentiation in the diagnosis of epithelial tumors.
R Moll
Sub-cellular biochemistry, 31, 205-262 (1999-02-05)
Ichiro Kurokawa et al.
Experimental dermatology, 20(3), 217-228 (2011-02-18)
Keratins are a highly diverse family of cytoskeletal proteins and important markers of epithelial cell differentiation. In this review, applying the new keratin nomenclature recently introduced, we summarize and discuss the distribution and significance of keratin patterns in cutaneous epithelial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service